Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ867027.1 | - | 18 | 4159 | ||
DQ867029.1 | - | 17 | 4051 | |||
DQ867030.1 | - | 16 | 3997 |
Gene Symbol: 5530400K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017425.1 | - | 1 | 1117 |
Gene Symbol: 6030403N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077881.1 | - | 1 | 657 |
Gene Symbol: A330076C08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034101.1 | - | 1938 |
Gene Symbol: Dynlt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166629.2 | NP_001160101.1 | 5 | 2376 | ||
GenBank mRNA | AK030569.1 | - | 5 | 2338 |
Gene Symbol: Dynlt1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014409.1 | - | 3 | 3012 |
Gene Symbol: Fbxl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015794.1 | NP_056609.1 | 9 | 9747 | ||
GenBank mRNA | AK018254.1 | - | 1 | 791 | ||
AK086279.1 | - | 1 | 1624 | |||
AK160716.1 | - | 1 | 385 |
Gene Symbol: Gm10789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141820.1 | - | 1 | 4241 |
Gene Symbol: Gm14340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133152.1 | - | 2 | 1575 |
Gene Symbol: Gm35387 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC066010.1 | - | 2 | 1680 |
Gene Symbol: Gm7969 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139503.1 | - | 3 | 2388 |
Gene Symbol: Hsd17b13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198030.2 | NP_932147.2 | 7 | 1587 | ||
GenBank mRNA | BC019427.1 | - | 7 | 1563 | 5132342 |
Gene Symbol: Oas1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB067529.1 | - | 6 | 1691 | ||
AF418004.1 | - | 6 | 1692 | |||
AK080152.1 | - | 6 | 1685 |
Gene Symbol: Zkscan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141505.1 | - | 1 | 1992 | ||
AK220177.1 | - | 5 | 4974 |