Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243046.1 | NP_001229975.1 | 15 | 1534 | ||
NM_001243047.1 | NP_001229976.1 | 11 | 1217 | |||
NM_011807.3 | NP_035937.2 | 23 | 2693 | |||
GenBank mRNA | AF388675.1 | - | 23 | 2472 | ||
AK030221.1 | - | 15 | 1536 | |||
AK039754.1 | - | 11 | 1082 |