Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199003.1 | NP_001185932.1 | 15 | 1485 | ||
NM_011880.3 | NP_036010.2 | 15 | 1485 | |||
GenBank mRNA | AF011360.1 | - | 15 | 1330 | ||
BC051133.1 | - | 15 | 1440 | 6588901 | ||
DQ139285.1 | - | 2 | 203 | |||
DQ139286.1 | - | 2 | 203 | |||
DQ139288.1 | - | 3 | 263 | |||
DQ139289.1 | - | 2 | 203 |