Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF025653.1 | - | 1, 2 | 103 | ||
AF034568.1 | - | 1, 2 | 157 | |||
AK002922.1 | - | 1, 2 | 206 | |||
AK079488.1 | - | 1, 2 | 203 | |||
AK142016.1 | - | 1, 2 | 200 | |||
AK150497.1 | - | 1, 2 | 210 | |||
AK151463.1 | - | 1, 2 | 224 | |||
AK160007.1 | - | 1, 2 | 212 | |||
AK160706.1 | - | 1, 2 | 190 | |||
AK169305.1 | - | 1, 2 | 176 | |||
BC043657.1 | - | 1, 2 | 175 | 5343539 |