Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011985.2 | NP_036115.1 | 1, 2 | 365 | ||
GenBank mRNA | AF085742.1 | - | 1, 2 | 365 | ||
AK146510.1 | - | 1, 2 | 172 | |||
BC107357.1 | - | 1, 2 | 272 | 40055939 | ||
BC107358.1 | - | 1, 2 | 272 | 40055946 |