Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011910.2 | NP_036040.1 | 3, 4 | 302 | ||
GenBank mRNA | AF172175.1 | - | 3, 4 | 302 | ||
AY065984.1 | - | 3, 4 | 266 | |||
BC107299.1 | - | 3, 4 | 290 | 40053156 | ||
BC107300.1 | - | 3, 4 | 288 | 40053161 |