Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB221554.1 | - | 2 | 407 | ||
AF149820.1 | - | 2 | 401 | |||
AF177755.1 | - | 2 | 389 | |||
AF177756.1 | - | 2 | 374 | |||
AF177757.1 | - | 2 | 1247 | |||
AK035490.1 | - | 2 | 404 | |||
AK076482.1 | - | 2 | 297 | |||
AK141466.1 | - | 2 | 297 | |||
AK145553.1 | - | 2 | 299 | |||
AK157424.1 | - | 2 | 396 | |||
BC016573.1 | - | 2 | 789 | |||
BC147770.1 | - | 2 | 499 | 9087493 | ||
BC147784.1 | - | 2 | 499 | 9087503 | ||
CK624100.1 | - | 2 | 285 | |||
DV655694.1 | - | 2 | 367 |