Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011929.2 | NP_036059.1 | 3 | 197 | ||
GenBank mRNA | AK145053.1 | - | 3 | 242 | ||
AK165362.1 | - | 3 | 243 |