Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011909.2 | NP_036039.2 | 8, 9 | 1145 | ||
GenBank mRNA | AF069502.1 | - | 8, 9 | 1106 | ||
AK004296.1 | - | 8, 9 | 1111 | |||
AK149772.1 | - | 8, 9 | 1146 | |||
AK151011.1 | - | 8, 9 | 1146 | |||
AK151260.1 | - | 8, 9 | 1146 | |||
AK151722.1 | - | 8, 9 | 1146 | |||
AK152034.1 | - | 8, 9 | 1111 | |||
AK152574.1 | - | 8, 9 | 1111 | |||
AK153312.1 | - | 8, 9 | 1115 | |||
BC138562.1 | - | 8, 9 | 1027 | 8861582 | ||
BC138577.1 | - | 8, 9 | 1027 | 8861597 | ||
CT010228.1 | - | 7, 8 | 881 |