Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243792.1 | NP_001230721.1 | 3, 4 | 712 | ||
NM_011935.3 | NP_036065.1 | 2, 3 | 650 | |||
GenBank mRNA | AB093266.1 | - | 3, 4 | 731 | ||
AB221580.1 | - | 2, 3 | 484 | |||
AF117254.1 | - | 2, 3 | 547 | |||
AK052731.1 | - | 3, 4 | 691 | |||
BC082324.1 | - | 2, 3 | 618 | 6850748 |