Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012001.2 | NP_036131.1 | 3 | 422 | ||
GenBank mRNA | AF071314.1 | - | 3 | 291 | ||
AK003752.1 | - | 3 | 284 | |||
AK009955.1 | - | 3 | 294 | |||
AK162763.1 | - | 3 | 288 | |||
AK167833.1 | - | 3 | 308 | |||
BC116800.1 | - | 3 | 276 | 40126119 | ||
BC116826.1 | - | 3 | 276 | 40126145 | ||
CX203220.1 | - | 3 | 422 |