Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012048.2 | NP_036178.1 | 2 | 279 | ||
GenBank mRNA | AB027563.1 | - | 2 | 267 | ||
AF163571.1 | - | 2 | 279 | |||
AK042272.1 | - | 2 | 374 | |||
AK138299.1 | - | 2 | 282 | |||
AY769919.1 | - | 1 | 93 | |||
AY769920.1 | - | 1 | 93 | |||
AY769921.1 | - | 1 | 93 | |||
AY769922.1 | - | 1 | 93 | |||
AY769923.1 | - | 1 | 93 | |||
AY769924.1 | - | 1 | 93 | |||
AY769925.1 | - | 1 | 93 | |||
AY769926.1 | - | 1 | 93 | |||
AY769927.1 | - | 1 | 93 | |||
AY769928.1 | - | 1 | 93 | |||
BC052820.1 | - | 2 | 352 | 30057338 |