Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013484.2 | NP_038512.2 | 2 | 328 | ||
GenBank mRNA | AK146812.1 | - | 2 | 329 | ||
BC011086.1 | - | 2 | 291 | 3992881 | ||
M57891.1 | - | 2 | 275 |