Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012010.3 | NP_036140.1 | 9 | 945 | ||
GenBank mRNA | AJ006587.1 | - | 9 | 935 | ||
AK010819.1 | - | 6 | 623 | |||
AK128949.1 | - | 9 | 944 | |||
AK146900.1 | - | 9 | 986 | |||
AK150095.1 | - | 9 | 952 | |||
AK151579.1 | - | 5 | 501 | |||
AK153568.1 | - | 5 | 501 | |||
AK165866.1 | - | 9 | 947 | |||
AK171317.1 | - | 9 | 946 | |||
BC036993.1 | - | 9 | 943 | |||
BC063755.1 | - | 9 | 945 | 6513251 | ||
BC139234.1 | - | 9 | 945 | 8862256 | ||
BC139235.1 | - | 9 | 945 | 8862257 |
Gene Symbol: Eif2s3x-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076900.1 | - | 1355 |