Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253808.1 | NP_001240737.1 | 7, 8 | 795 | ||
NM_001253809.1 | NP_001240738.1 | 7, 8 | 791 | |||
NM_012025.7 | NP_036155.1 | 8, 9 | 926 | |||
GenBank mRNA | AB030252.1 | - | 7, 8 | 775 | ||
AF079974.1 | - | 7, 8 | 736 | |||
AF212320.1 | - | 7, 8 | 762 | |||
AF212321.1 | - | 8, 9 | 893 | |||
AK052423.1 | - | 1 | 1781 | |||
AK053850.1 | - | 7, 8 | 756 | |||
AK144608.1 | - | 8, 9 | 894 | |||
AK144745.1 | - | 7, 8 | 755 | |||
AK154929.1 | - | 8, 9 | 914 | |||
AK162660.1 | - | 5, 6 | 3995 | |||
AK165897.1 | - | 7, 8 | 748 | |||
AK166084.1 | - | 8, 9 | 884 | |||
AK166094.1 | - | 7, 8 | 753 | |||
AK168020.1 | - | 7, 8 | 752 | |||
AK168025.1 | - | 7, 8 | 752 | |||
AK168170.1 | - | 7, 8 | 752 | |||
AK168679.1 | - | 7, 8 | 736 | |||
AK181912.1 | - | 4, 5 | 320 | |||
AK183940.1 | - | 5, 6 | 386 | |||
AK184356.1 | - | 3, 4 | 225 | |||
AK193352.1 | - | 3, 4 | 225 | |||
AK193637.1 | - | 3, 4 | 225 | |||
AK200143.1 | - | 3, 4 | 225 | |||
AK207314.1 | - | 5, 6 | 386 | |||
BC010715.1 | - | 7, 8 | 741 | 3602242 |