Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Add2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271857.1 | NP_001258786.1 | 14 | 2131 | ||
NM_001271858.1 | NP_001258787.1 | 14 | 2128 | |||
NM_001271859.1 | NP_001258788.1 | 14 | 1929 | |||
NM_013458.5 | NP_038486.2 | 14 | 1932 | |||
GenBank mRNA | AF100422.1 | - | 13 | 1807 | ||
AF189769.1 | - | 12 | 1729 | |||
AF189770.1 | - | 7 | 985 | |||
AK014496.1 | - | 14 | 1897 | |||
AK029488.1 | - | 14 | 2082 | |||
AK147544.1 | - | 14 | 2009 | |||
AK156954.1 | - | 14 | 1921 |