Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013519.2 | NP_038547.2 | 1 | 962 | ||
GenBank mRNA | AK076319.1 | - | 1 | 963 | ||
AK146887.1 | - | 1 | 963 | |||
BC139208.1 | - | 1 | 605 | 8862230 | ||
S63607.1 | - | 1 | 480 | |||
X74040.1 | - | 1 | 939 | |||
X92499.1 | - | 1 | 546 |