Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161775.1 | NP_001155247.1 | 2 | 361 | ||
NM_013607.2 | NP_038635.2 | 2 | 361 | |||
GenBank mRNA | AK085742.1 | - | 2 | 319 | ||
AK137226.1 | - | 2 | 318 | |||
AK137716.1 | - | 2 | 317 | |||
AK173045.1 | - | 2 | 325 | |||
BC026142.1 | - | 2 | 346 | 3989495 | ||
D85923.1 | - | 2 | 316 | |||
D85924.1 | - | 2 | 316 | |||
L25860.1 | - | 2 | 305 | |||
S81516.1 | - | 2 | 288 |