Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hapln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013500.4 | NP_038528.3 | 3 | 532 | ||
GenBank mRNA | AF139572.1 | - | 1340 | |||
AK003582.1 | - | 3 | 311 | |||
AK032750.1 | - | 3 | 533 | |||
AK048107.1 | - | 3 | 316 | |||
BC066853.1 | - | 4 | 379 | 30430221 |