Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013541.1 | NP_038569.1 | 3 | 181 | ||
GenBank mRNA | AK079144.1 | - | 1 | 109 | ||
BC002048.1 | - | 3 | 186 | 3593257 | ||
BC061109.1 | - | 3 | 190 | 30322194 | ||
D30687.1 | - | 3 | 181 | |||
X53451.1 | - | 2 | 140 |
Gene Symbol: Gstp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181796.2 | NP_861461.1 | 3 | 188 | ||
GenBank mRNA | BC064781.1 | - | 3 | 188 | 30112237 | |
BC094623.1 | - | 3 | 192 | 1381023 | ||
BC100587.1 | - | 3 | 205 | |||
GQ181117.1 | - | 1 | 98 |