Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013557.2 | NP_038585.2 | 493 | |||
GenBank mRNA | AK014775.1 | - | 455 | |||
AK020887.1 | - | 339 | ||||
AK155206.1 | - | 475 | ||||
AK156492.1 | - | 454 | ||||
AK158437.1 | - | 449 | ||||
AK173160.1 | - | 443 | ||||
AY033898.1 | - | 478 | ||||
BB625879.1 | - | 494 | ||||
BC028923.1 | - | 465 | 4164417 | |||
BC111035.1 | - | 444 | 5357121 |