Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013562.2 | NP_038590.2 | 3 | 454 | ||
GenBank mRNA | AK077790.1 | - | 3 | 450 | ||
AK078888.1 | - | 451 | ||||
AK168682.1 | - | 3 | 451 | |||
BC037013.1 | - | 3 | 428 | |||
BC043723.1 | - | 3 | 421 | 5369100 | ||
X17400.1 | - | 3 | 431 |