Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mc5r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013596.2 | NP_038624.2 | 2 | 857 | ||
GenBank mRNA | BC100720.1 | - | 2 | 854 | 40045173 | |
BC100721.1 | - | 2 | 854 | 40045171 | ||
BC100722.1 | - | 2 | 854 | 40045170 | ||
BC100723.1 | - | 2 | 854 | 40045167 | ||
U08354.1 | - | 2 | 863 |
Set Membership: |
G-protein coupled receptor |