Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123037.2 | NP_001116509.1 | 10 | 1303 | ||
NM_001123038.2 | NP_001116510.1 | 11 | 2259 | |||
NM_013506.3 | NP_038534.2 | 11 | 1443 | |||
GenBank mRNA | AK076509.1 | - | 11 | 1154 | ||
AK088669.1 | - | 11 | 1971 | |||
AK165898.1 | - | 10 | 1014 | |||
BC094422.1 | - | 11 | 1139 | 6808404 | ||
CD773468.1 | - | 1 | 399 | |||
X12507.1 | - | 11 | 1155 |