Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013594.2 | NP_038622.2 | 15 | 2652 | ||
GenBank mRNA | AF072240.1 | - | 17 | 2799 | ||
AK004624.2 | - | 15 | 2653 | |||
AK156401.1 | - | 16 | 2643 | |||
BC069837.1 | - | 15 | 2668 | 30435760 |