Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nodal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013611.4 | NP_038639.2 | 3 | 1459 | ||
GenBank mRNA | AK049290.1 | - | 3 | 1143 | ||
AK082823.1 | - | 3 | 1143 | |||
X70514.1 | - | 3 | 1458 |