Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013624.2 | NP_038652.2 | 3 | 236 | ||
GenBank mRNA | AK165831.1 | - | 3 | 221 | ||
U96411.1 | - | 3 | 236 |