Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013678.2 | NP_038706.1 | 1, 2 | 100 | ||
GenBank mRNA | AK157125.1 | - | 1, 2 | 114 | ||
AK165559.1 | - | 1, 2 | 107 | |||
AK166181.1 | - | 1, 2 | 129 | |||
AK172018.1 | - | 1, 2 | 103 | |||
AK216211.1 | - | 14 | ||||
BC049779.1 | - | 1, 2 | 91 | |||
BC096401.1 | - | 1, 2 | 100 | 1365532 |