Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D13738.1 | - | 2 | 423 | ||
S67051.1 | - | 3 | 87 | |||
X71426.1 | - | 3 | 158 |