Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013721.3 | NP_038749.1 | 3 | 158 | ||
GenBank mRNA | AK019449.1 | - | 3 | 161 | ||
AK135447.1 | - | 3 | 164 | |||
AK160044.1 | - | 3 | 160 | |||
AK160621.1 | - | 3 | 159 | |||
AK160622.1 | - | 3 | 166 | |||
BC029992.1 | - | 2 | 147 | |||
BC052339.1 | - | 2 | 151 | |||
BC065176.1 | - | 2 | 146 | 6819288 | ||
BC080663.1 | - | 2 | 147 | 4487611 | ||
BC080669.1 | - | 2 | 147 | 5060194 | ||
BC080712.1 | - | 2 | 142 | 4457484 | ||
BC084678.1 | - | 2 | 159 | 30431224 | ||
BC089624.1 | - | 3 | 163 | 6763528 | ||
BC091731.1 | - | 2 | 142 | 2655142 | ||
BC091769.1 | - | 2 | 147 | 4158894 | ||
BC099389.1 | - | 2 | 142 | 30526878 |