Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013755.3 | NP_038783.1 | 2 | 246 | ||
GenBank mRNA | AA553085.1 | - | 2 | 245 | ||
AF114031.1 | - | 2 | 181 | |||
AK159576.1 | - | 2 | 227 | |||
AK202350.1 | - | 2 | 144 | |||
AK205608.1 | - | 2 | 144 | |||
AK209105.1 | - | 2 | 163 | |||
AK209409.1 | - | 2 | 144 | |||
BC029903.1 | - | 2 | 207 | 4487769 |