Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013739.2 | NP_038767.1 | 5 | 1417 | ||
GenBank mRNA | AF179242.1 | - | 5 | 1417 | ||
AF237580.1 | - | 5 | 1413 | |||
AK137520.1 | - | 5 | 1408 | |||
AK154750.1 | - | 4 | 2240 | |||
AK155180.1 | - | 4 | 2249 | |||
AK156060.1 | - | 4 | 2247 | |||
AK172511.1 | - | 4 | 2247 | |||
BC048784.1 | - | 4 | 2548 | |||
BC116922.1 | - | 5 | 1416 | 40126241 | ||
BC120535.1 | - | 5 | 1416 | 40129458 |