Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163182.1 | NP_001156654.1 | 8 | 1485 | ||
NM_001163183.1 | NP_001156655.1 | 8 | 1443 | |||
NM_001163184.1 | NP_001156656.1 | 7 | 1338 | |||
NM_001163185.1 | NP_001156657.1 | 7 | 1328 | |||
NM_001163186.1 | NP_001156658.1 | 7 | 1434 | |||
NM_001163187.1 | NP_001156659.1 | 6 | 1329 | |||
NM_001291336.1 | NP_001278265.1 | 6 | 1298 | |||
NM_013746.3 | NP_038774.1 | 9 | 1590 | |||
GenBank mRNA | AF000272.1 | - | 9 | 1561 | ||
AF071000.1 | - | 6 | 1317 | |||
AF100613.1 | - | 7 | 1422 | |||
AF101053.1 | - | 8 | 1456 | |||
AK005102.1 | - | 7 | 1293 | |||
AK141634.1 | - | 1297 | ||||
AK164683.1 | - | 7 | 1293 | |||
BC024756.1 | - | 9 | 1551 | 5369461 |