Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013765.2 | NP_038793.2 | 4 | 409 | ||
GenBank mRNA | AK008301.1 | - | 4 | 405 | ||
AK010689.2 | - | 4 | 404 | |||
AK018702.1 | - | 4 | 405 | |||
AK152132.1 | - | 4 | 405 | |||
AV585706.1 | - | 3 | 363 | |||
BC036987.1 | - | 3 | 395 | 4987659 | ||
BC071192.1 | - | 1249 | ||||
BC081452.1 | - | 3 | 382 | 6411109 | ||
BC100456.1 | - | 4 | 396 | 6541446 | ||
BU530648.1 | - | 4 | 410 | |||
U67770.1 | - | 3 | 386 |