Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013783.2 | NP_038811.2 | 15, 16 | 1794 | ||
GenBank mRNA | AF157105.1 | - | 15, 16 | 1740 | ||
AF157106.1 | - | 13, 14 | 1449 | |||
AF176569.1 | - | 15, 16 | 1791 | |||
AF302075.1 | - | 13, 14 | 1415 | |||
AF302076.1 | - | 14, 15 | 1484 | |||
AF302077.1 | - | 14, 15 | 1526 | |||
AK154366.1 | - | 12, 13 | 1283 | |||
BC137649.1 | - | 13, 14 | 1567 | 8860668 | ||
BC144804.1 | - | 13, 14 | 1567 | 9053352 |