Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ror2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013846.3 | NP_038874.3 | 1, 2 | 337 | ||
GenBank mRNA | AB010384.1 | - | 1, 2 | 105 | ||
AK031112.1 | - | 1, 2 | 307 | |||
AK031789.1 | - | 1, 2 | 95 | |||
AK083538.1 | - | 1, 2 | 313 | |||
AK084752.1 | - | 1, 2 | 303 | |||
AK157705.1 | - | 1, 2 | 353 | |||
BC070436.1 | - | 1, 2 | 337 | 30535615 |