Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013881.4 | NP_038909.3 | 2 | 629 | ||
GenBank mRNA | AB019577.1 | - | 2 | 154 | ||
AF145922.1 | - | 2 | 522 | |||
AK122331.2 | - | 2 | 611 | |||
AK146620.1 | - | 2 | 591 | |||
AK148142.1 | - | 2 | 630 | |||
AK208350.1 | - | 1 | 29 | |||
BC013691.1 | - | 2 | 39 | |||
BC046778.1 | - | 2 | 587 | 5709559 | ||
BC053029.1 | - | 2 | 587 | 5709559 |