Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013923.2 | NP_038951.1 | 2 | 500 | ||
GenBank mRNA | AF120206.1 | - | 393 | |||
AF120207.1 | - | 1 | 393 | |||
AK082665.1 | - | 2 | 452 | |||
AK147768.1 | - | 2 | 488 | |||
AK166197.1 | - | 2 | 488 | |||
BC040769.1 | - | 1 | 275 | 4457807 |