Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013876.3 | NP_038904.1 | 3 | 1765 | ||
GenBank mRNA | AB024427.1 | - | 4 | 1630 | ||
AK003552.1 | - | 1 | 90 | |||
AK014254.1 | - | 3 | 1919 | |||
AK151082.1 | - | 607 | ||||
AK153482.1 | - | 608 | ||||
BC010299.1 | - | 3 | 1765 | 3155804 |