Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cngb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013927.2 | NP_038955.1 | 17 | 2147 | ||
GenBank mRNA | AJ243572.1 | - | 17 | 2147 | ||
AK040140.1 | - | 8 | 1014 |