Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282428.1 | NP_001269357.1 | 3 | 654 | ||
NM_002252.4 | NP_002243.3 | 3 | 604 | |||
GenBank mRNA | AF043472.1 | - | 2 | 334 | ||
AK225833.1 | - | 3 | 654 | |||
AK314451.1 | - | 3 | 604 | |||
BC004148.2 | - | 3 | 541 | 2958834 | ||
BC004987.1 | - | 3 | 541 | 2957877 | ||
BC015947.2 | - | 3 | 593 | 3919594 |