Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015773.2 | NP_056588.1 | 2, 3 | 273 | ||
GenBank mRNA | AF173866.1 | - | 2, 3 | 210 | ||
AK005732.1 | - | 2, 3 | 221 | |||
AK054199.1 | - | 1, 2 | 309 | |||
BC090660.1 | - | 2, 3 | 216 | 30673350 | ||
BC100455.1 | - | 2, 3 | 223 | 30919921 | ||
CJ060006.1 | - | 2, 3 | 273 |