Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icosl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015790.3 | NP_056605.1 | 3 | 457 | ||
GenBank mRNA | AF199027.1 | - | 3 | 366 | ||
AF216747.1 | - | 2 | 300 | |||
AF394451.1 | - | 300 | ||||
AK041320.1 | - | 2 | 925 | |||
AK041421.1 | - | 3 | 467 | |||
BC029227.1 | - | 3 | 457 | 4217333 |