Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016662.4 | NP_057871.2 | 4, 5 | 379 | ||
GenBank mRNA | AK086190.1 | - | 2, 3 | 1135 | ||
AK138742.1 | - | 4, 5 | 380 | |||
BC051970.1 | - | 4, 5 | 382 | 5714980 | ||
U32394.1 | - | 4, 5 | 316 |