Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130458.1 | NP_001123930.1 | 4 | 582 | ||
NM_001130459.1 | NP_001123931.1 | 4 | 551 | |||
NM_015749.3 | NP_056564.1 | 4 | 585 | |||
GenBank mRNA | AF090686.1 | - | 3 | 512 | ||
AK133707.1 | - | 4 | 547 | |||
AK143761.1 | - | 4 | 553 | |||
AK147176.1 | - | 4 | 556 | |||
AK161618.1 | - | 4 | 552 | |||
AK170401.1 | - | 4 | 571 | |||
BC003720.1 | - | 3 | 458 | 3601531 |