Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015780.2 | NP_056595.1 | 2, 3 | 341 | ||
GenBank mRNA | BC111870.1 | - | 2, 3 | 324 | 40057075 | |
M29008.1 | - | 2, 3 | 341 |