Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261408.1 | NP_001248337.1 | 4 | 466 | ||
NM_001261409.1 | NP_001248338.1 | 2 | 452 | |||
NM_003567.3 | NP_003558.1 | 2 | 429 | |||
XM_017002480.1 | XP_016857969.1 | 4 | 842 | |||
GenBank mRNA | AF124250.1 | - | 2 | 421 | ||
AK055442.1 | - | 2 | 452 | |||
BC028477.1 | - | 4 | 495 | |||
BC039895.1 | - | 2 | 322 | 6060541 | ||
CN337133.1 | - | 2 | 431 | |||
U92715.1 | - | 2 | 237 |