Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015791.3 | NP_056606.2 | 2 | 404 | ||
GenBank mRNA | AF176527.1 | - | 2 | 411 | ||
AK002538.1 | - | 2 | 415 | |||
AK012401.1 | - | 2 | 422 | |||
AK029989.1 | - | 2 | 405 | |||
AK077595.1 | - | 2 | 426 | |||
AK078111.1 | - | 2 | 424 | |||
AK147748.1 | - | 2 | 421 | |||
AK160145.1 | - | 2 | 385 | |||
AK162592.1 | - | 2 | 421 | |||
BC011100.1 | - | 2 | 425 | 4011855 | ||
BC034854.1 | - | 2 | 336 | 3468704 |