Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163704.1 | NP_001157176.1 | 2 | 550 | ||
NM_001163705.1 | NP_001157177.1 | 2 | 334 | |||
NM_001163706.1 | NP_001157178.1 | 2 | 326 | |||
NM_001163707.1 | NP_001157179.1 | 2 | 361 | |||
NM_015797.4 | NP_056612.1 | 2 | 1076 | |||
GenBank mRNA | AF176526.1 | - | 2 | 449 | ||
AK002840.1 | - | 2 | 288 | |||
AK144715.1 | - | 2 | 487 | |||
AK150612.1 | - | 2 | 518 | |||
AK152888.1 | - | 2 | 400 | |||
AK165867.1 | - | 2 | 1061 | |||
AK170743.1 | - | 2 | 327 | |||
AK171483.1 | - | 2 | 327 | |||
BB854497.1 | - | 2 | 350 | |||
BC017512.1 | - | 2 | 346 | 4485218 | ||
BY260050.1 | - | 2 | 318 |