Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016659.3 | NP_057868.2 | 7 | 1090 | ||
GenBank mRNA | AF074456.1 | - | 5 | 708 | ||
AF218077.1 | - | 4 | 561 | |||
AF374001.1 | - | 5 | 596 | |||
AF374002.1 | - | 5 | 596 | |||
AK080158.1 | - | 5 | 672 | |||
AY078436.1 | - | 7 | 1090 | |||
BC140968.1 | - | 4 | 566 | 9055580 | ||
BC144929.1 | - | 3 | 476 | 9053477 | ||
M25775.1 | - | 5 | 694 | |||
M25812.1 | - | 5 | 628 | |||
U34890.1 | - | 4 | 514 |